Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDK3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552432
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552432 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552432 Supplier Invitrogen™ Supplier No. PA552432
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSEDTWPGVT QLPDYKGSFP KWTRKGLEEI VPNLEPEGRD LLMQLLQYDP SQRITAKTAL AHPYFSSPEP SPAARQYVLQ RFRH Highest antigen sequence identity to the following orthologs: Mouse - 49%, Rat - 49%.

CDK3/CyclinE1is a member of the cyclin-dependent protein kinase family and promotes entry into S phase, in part by activating members of the E2F family of transcription factors. CDK3 can also associate with cyclin C and phosphorylates the retinoblastoma 1 protein to promote exit from G0. The CDK3 gene has been mapped to chromosomal location of 17q22-qter, telomeric to the BRCA1 gene by somatic cell hybrids analysis. The presence of a single point mutation in the CDK3 gene from several Mus musculus strains commonly used in the laboratory has been reported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDK3
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Cdk3
Gene Accession No. Q00526
Gene Alias 2310050O10Rik; Cdk3; Cdk3-ps; CDKN3; Cell division protein kinase 3; cyclin dependent kinase 3; cyclin-dependent kinase 3; cyclin-dependent kinase 3, pseudogene
Gene Symbols Cdk3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CDK3. Recombinant protein control fragment (Product #RP-89774).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1018
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.