Learn More
TARGETMOL CHEMICALS INC CDK4/6/1 INHIBITOR 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T10735
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. CDK4/6/1 Inhibitor (Crozbaciclib) is a type of CDK4/6 inhibitor (IC50s 3 and 1 nM). CDK4/6 inhibitor is a class of compounds used for the treatment of some types of hormone receptor positive HER2-negative breast cancer which can block the process of breast cancer cell division and reproduction. purity: 99%
Catalog No. 50-258-1679
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.