Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDK6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. p-7205699 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-9176 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-9176 Supplier Proteintech Group Inc Supplier No. 140521AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IARS2 belongs to the class-I aminoacyl-tRNA synthetase family. Members of class I have two highly conserved sequence motifs. They aminoacylate at the 2′-OH of an adenosine nucleotide, and are usually monomeric or dimeric (one or two subunits, respectively). Both classes of aminoacyl-tRNA synthetases are multidomain proteins. The catalytic domains of all the aaRSs of a given class are found to be homologous to one another, while class I and class II aaRSs are unrelated to one another. The class I aaRSs have the ubiquitous Rossmann fold and have the antiparallel beta-strands architecture while the class II aaRSs have a unique fold made up of antiparallel beta-strands.

Specifications

Antigen CDK6
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.17 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CDK6
Gene Accession No. Q00534, Q64261
Gene Alias CDK6, Cell division protein kinase 6, cyclin dependent kinase 6, PLSTIRE
Gene Symbols Cdk6
Host Species Rabbit
Immunogen CDK6 Fusion Protein Ag5275
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1021, 114483, 12571
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.