Learn More
TARGETMOL CHEMICALS INC CDK9 INHIBITOR HH1 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T118066
Encompass_Preferred
Also available in 1mg 10mg 25mg 50mg 100mg 500mg and bulk. Please contact Fisher for quotes. CDK9 inhibitor HH1 (8019-9719) is an inhibitor of the human CDK2-cyclin A2 complex with an IC50 value of 2 μM. purity: 97%
Catalog No. 50-257-7444
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.