Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDKN3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550929
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550929 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550929 Supplier Invitrogen™ Supplier No. PA550929
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CDKN3 Polyclonal Antibody detects endogenous levels of CDKN3 protein.

The protein encoded by this gene belongs to the dual specificity protein phosphatase family. It was identified as a cyclin-dependent kinase inhibitor, and has been shown to interact with, and dephosphorylate CDK2 kinase, thus prevent the activation of CDK2 kinase. This gene was reported to be deleted, mutated, or overexpressed in several kinds of cancers. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDKN3
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.02% sodium azide; pH 7.4
Gene CDKN3
Gene Accession No. Q16667, Q810P3
Gene Alias 2410006H10Rik; CDI1; CDK2-associated dual specificity phosphatase; CDK2-associated dual-specificity phosphatase; CDK2-associated dual-specificity phosphatase {ECO:0000250; Cdk-associated protein phosphatase; Cdkn3; cdkn3 {ECO:0000312; CIP2; cyclin dependent kinase inhibitor 3; cyclin-dependent kinase inhibitor; cyclin-dependent kinase inhibitor 3; cyclin-dependent kinase inhibitor 3 {ECO:0000250; cyclin-dependent kinase interacting protein 2; cyclin-dependent kinase interactor 1; Cyclin-dependent kinase-interacting protein 2; EMBL:AAI67026.1, ECO:0000312; KAP; KAP1; kinase associated phosphatase; kinase-associated phosphatase; kinase-associated phosphatase {ECO:0000250; RGD:1307781}; UniProtKB:Q16667}
Gene Symbols CDKN3
Host Species Rabbit
Immunogen Synthesized peptide derived from the Internal region of human CDKN3.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1033, 72391
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.