Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDR2L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112378
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112378 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112378 Supplier Invitrogen™ Supplier No. PA5112378
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CDR2L Polyclonal Antibody for Western Blot, IHC (P), ELISA

The function of CDR2L remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDR2L
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 3.57 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Cdr2l
Gene Accession No. Q86X02
Gene Alias CDR2L; cerebellar degeneration related protein 2 like; cerebellar degeneration-related protein 2-like; D030068L24Rik; Gm21; HUMPPA; Paraneoplastic 62 kDa antigen; paraneoplastic antigen; RGD1306881
Gene Symbols Cdr2l
Host Species Rabbit
Immunogen Recombinant Human Cerebellar degeneration-related protein 2-like protein (376-465AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30850
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.