Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDSN Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. p-7205452 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-8552 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-8552 Supplier Proteintech Group Inc Supplier No. 131841AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CDSN, also named as S protein, is important for the epidermal barrier integrity. CDSN is a unique extracellular 52- to 56- kDa glycoprotein found in corneodesmosomes. Pathogenic mutations in CDSN have been linked in humans to genodermatoses affecting both the hair follicle and the epidermis. Inflammatory peeling skin syndrome is caused a novel mutation in CDSN.

Specifications

Antigen CDSN
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.19 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CDSN
Gene Accession No. Q15517, Q7TPC1
Gene Alias CDSN, corneodesmosin, D6S586E, HTSS, S
Gene Symbols Cdsn
Host Species Rabbit
Immunogen CDSN Fusion Protein Ag3854
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1041, 386463
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.