Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542861
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542861 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542861 Supplier Invitrogen™ Supplier No. PA542861
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: PATPPATPPA ASPSALKGVS QDLLERIRAK EAQKQLAQMT RCPEQEQRLQ Sequence homology: Cow: 93%; Dog: 77%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 93%; Rat: 100%; Yeast: 85%.

KMT2A is a histone methyltransferase that plays an essential role in early development and hematopoiesis. It is a catalytic subunit of the MLL1/MLL complex, a multiprotein complex that mediates both methylation of 'Lys-4' of histone H3 (H3K4me) complex and acetylation of 'Lys-16' of histone H4 (H4K16ac). In the MLL1/MLL complex, KMT2A specifically mediates H3K4me, a tag for epigenetic transcriptional activation. KMT2A has weak methyltransferase activity by itself, and requires other components of the MLL1/MLL complex to obtain full methyltransferase activity. It has no activity toward histone H3 phosphorylated on 'Thr-3', less activity toward H3 dimethylated on 'Arg-8' or 'Lys-9', and has higher activity toward H3 acetylated on 'Lys-9'. Required for transcriptional activation of HOXA9 and promotes PPP1R15A-induced apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDT1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CDT1
Gene Accession No. Q8R4E9, Q9H211
Gene Alias 2610318F11Rik; AW545653; C76791; CDT1; chromatin licensing and DNA replication factor 1; DNA replication factor Cdt1; Double parked homolog; Double parked, Drosophila, homolog of; DUP; Retroviral insertion site 2 protein; retroviral integration site 1; retroviral integration site 2; RIS2
Gene Symbols CDT1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human CDT1 (aa 400-449).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 292071, 67177, 81620
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.