Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDX1 Monoclonal Antibody (OTI2A5)
GREENER_CHOICE

Catalog No. PIMA527317
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527317 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527317 Supplier Invitrogen™ Supplier No. MA527317
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CDX1 Monoclonal Antibody for Western Blot, IHC (P)

CDX1, or caudal type homeobox 1, is a gene encoding a homeodomain transcription factor that plays a significant role in the development and differentiation of intestinal epithelial cells. CDX1 is expressed in the intestinal crypts, where it is integral to the Wnt/B-catenin signaling pathway, crucial for cell proliferation. Interestingly, while CDX1 expression is initially active, its expression is often silenced as colon cancer progresses in humans, suggesting a complex role in tumor biology. The CDX1 protein functions as a transcriptional regulator that influences proliferation and differentiation, expressing early during intestinal development. In mice, it is involved in the development of the posterior embryo and is associated with abnormal differentiation in metaplastic and neoplastic tissues, providing insight into intestinal-type gastric adenocarcinoma progression. CDX1 is related to other vertebrate Cdx genes such as Cdx2 and Cdx4, which are homologues of the Drosophila caudal gene and are also implicated in embryonic development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDX1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2A5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CDX1
Gene Accession No. P47902
Gene Alias caudal type homeo box 1; caudal type homeo box transcription factor 1; caudal type homeobox 1; caudal type homeobox transcription factor 1; caudal-type homeobox protein 1; caudal-type homeobox protein CDX1; Cdx; CDX1; Cdx-1; Homeobox protein CDX-1
Gene Symbols CDX1
Host Species Mouse
Immunogen Full length human recombinant protein of CDX1 produced in E.coli.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1044
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.