Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDYL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117860
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117860 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117860 Supplier Invitrogen™ Supplier No. PA5117860
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CDYL Polyclonal Antibody for Western Blot, IHC (P), ICC/IF, IP

Chromodomain Y is a primate-specific Y-chromosomal gene family expressed exclusively in the testis and implicated in infertility. Although the Y-linked genes are testis-specific, this autosomal gene is ubiquitously expressed. The Y-linked genes arose by retrotransposition of an mRNA from this gene, followed by amplification of the retroposed gene. Proteins encoded by this gene superfamily possess a chromodomain, a motif implicated in chromatin binding and gene suppression, and a catalytic domain believed to be involved in histone acetylation. Multiple proteins are encoded by transcript variants of this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDYL
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene CDYL
Gene Accession No. Q9Y232
Gene Alias AI325931; CDYL; CDYL1; CDY-like; CDY-like, autosomal; chromodomain protein Y-like variant 2; chromodomain protein Y-like variant 3; chromodomain protein Y-like variant 4; chromodomain protein, Y chromosome-like; chromodomain protein, Y-like; chromodomain Y like; chromodomain Y-like protein; chromodomain Y-like protein pseudogene; Crotonyl-CoA hydratase; DKFZp586C1622; MGC131936; Putative histone acetyltransferase Cdyl; Putative tubulin acetyltransferase Cdyl; testis-specific chromodomain Y-like protein
Gene Symbols CDYL
Host Species Rabbit
Immunogen E. coli-derived Human CDYL fragment.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9425
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.