Learn More
Novus Biologicals Maurice CE-SDS PLUS 1x Sample Buffer, 25 mL
SDP
Supplier:  Novus Biologicals 046567
Encompass
Buffer for samples preparation for CE-SDS PLUS application, 25 mL. For use with CE-SDS PLUS cartridge (PS-MC02-SP), or CE-SDS cartridge (PS-MC02-S) on Maurice (090-000) and Maurice S. (090-001) systems.
Catalog No. NC1634279