Learn More
CELL MARQUE CORPORATION EA (CEA31) Mouse Monoclonal Antibody, 1.0 mL concentrate
SDP
Supplier:  CELL MARQUE CORPORATION 236M96
Encompass
Anti-CEA is an antibody against carcinoembryonic antigen (CEA), a protein thought to promote tumor development through its role as a cell adhesion molecule. Anti-CEA positivity is seen in adenocarcinomas of many origins, especially colon and lung adenocarcinomas, but rarely seen in mesothelial cells and mesotheliomas. 1-5
Catalog No. NC2873933
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.