Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ WNK4 Antibody Blocking Peptide
SDP

Catalog No. NB600284PEP Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600284PEP 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600284PEP Supplier Novus Biologicals™ Supplier No. NB600284PEP
Add to Cart
Add to Cart

Blocking peptide for NB600-284

Application Notes:This peptide is useful as a blocking peptide for NB600-284.

Specifications

Gene ID (Entrez) 65266
Species Human
Common Name WNK4
Concentration 1 mg/mL
Content And Storage Aliquot and store at -20 C or -80 C. Avoid freeze-thaw cycles.
Formulation PBS
For Use With (Application) This Peptide is Useful as a Blocking Peptide for NB600-284. For Further Blocking Peptide Related Protocol, Click Here .
Gene Alias EC 2.7.11, EC 2.7.11.1, PHA2B, PRKWNK4protein kinase, lysine deficient 4, Protein kinase lysine-deficient 4, Protein kinase with no lysine 4, serine/threonine-protein kinase WNK4, WNK lysine deficient protein kinase 4
Gene Symbol WNK4
Quantity 0.1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.