Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CEACAM19 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543365
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543365 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543365 Supplier Invitrogen™ Supplier No. PA543365
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MLLRRAQPTD SGTYQVAITI NSEWTMKAKT EVQVAEKNKE LPSTHLPTNA Sequence homology: Human: 100%.

The specific function of the protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEACAM19
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CEACAM19
Gene Accession No. Q7Z692
Gene Alias C130022P09Rik; carcinoembryonic antigen related cell adhesion molecule 19; carcinoembryonic antigen-like 1; Carcinoembryonic antigen-related cell adhesion molecule 19; CEACAM19; CEACM19; CEAL1; CEA-related cell adhesion molecule 19; UNQ2973/PRO7436
Gene Symbols CEACAM19
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human CEACAM19.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56971
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.