Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CEACAM6 Recombinant Rabbit Monoclonal Antibody (408), PE
GREENER_CHOICE

Catalog No. PIMA546697
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA546697 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA546697 Supplier Invitrogen™ Supplier No. MA546697
Only null left
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Preparation: This antibody was obtained from a rabbit immunized with purified, recombinant Human CD66c (CEACAM6) (Met1-Gly320) and conjugated with PE under optimum conditions, the unreacted PE was removed. Storage: This antibody can be stored at 2C-8C for twelve months without detectable loss of activity. Protected from prolonged exposure to light. Do not freeze! Sodium azide is toxic to cells and should be disposed of properly. Flush with large volumes of water during disposal.

CEA Cell Adhesion Molecule 6 (CEACAM6) belongs to the carcinoembryonic antigen (CEA) family whose members are glycosyl phosphatidyl inositol (GPI) anchored cell surface glycoproteins. Members of this family play a role in cell adhesion and are widely used as tumor markers in serum immunoassay determinations of carcinoma. CEACAM6 affects the sensitivity of tumor cells to adenovirus infection. The acts as a receptor for adherent-invasive E. coli adhesion to the surface of ileal epithelial cells in patients with Crohn's disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEACAM6
Applications Flow Cytometry
Classification Recombinant Monoclonal
Clone 408
Concentration 100 μg/mL
Conjugate PE
Formulation PBS with 0.5% BSA and 0.09% sodium azide
Gene CEACAM6
Gene Accession No. P40199
Gene Alias BGP; BGP-1; carcinoembryonic antigen related cell adhesion molecule 6; carcinoembryonic antigen-related cell adhesion molecule 6; carcinoembryonic antigen-related cell adhesion molecule 6 (non-specific cross reacting antigen); CD66a; CD66c; CD66d; CD66e; CEA; CEACAM-1; CEACAM-3; CEACAM-5; CEACAM6; CEACAM-6; CEAL; CGM1; Cluster of Differentiation 66c; NCA; Non-specific crossreacting antigen; Normal cross-reacting antigen; pan CEACAM
Gene Symbols CEACAM6
Host Species Rabbit
Immunogen Recombinant protein ecoding Human CEACAM6 (Met1-Gly320).
Purification Method Protein A
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4680
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.