Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEBPB Mouse anti-Human, Rat, Clone: 2B6E10, Proteintech
SDP

Catalog No. 501737067 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-7067 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-7067 Supplier Proteintech Group Inc Supplier No. 666491IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This gene belongs to the forkhead family of transcription factors which is characterized by a distinct DNA-binding forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in the development of mesenchymal tissues.

Specifications

Antigen CEBPB
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 2B6E10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene CEBPB
Gene Accession No. P17676, P21272
Gene Alias C/EBP beta, CEBPB, CRP2, IL6DBP, LAP, Liver activator protein, NF IL6, Nuclear factor NF IL6, TCF 5, TCF5, Transcription factor 5
Gene Symbols CEBPB
Host Species Mouse
Immunogen CEBPB Fusion Protein Ag20073
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1051, 24253
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.