Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CEDIA™ Benzodiazepine Assays

Catalog No. 21200438
Change view
Click to view available options
Quantity:
1 x 495 mL, LC
1 x 65 mL, MCC
3 x 17 mL, Indiko
3 x 17 mL, TSC
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-200-438 1 x 495 mL, LC
23-278-208 3 x 17 mL, TSC
23-278-206 3 x 17 mL, Indiko
23-278-207 1 x 65 mL, MCC
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 21-200-438 Supplier Thermo Scientific™ Supplier No. 1775561
Add to Cart
Add to Cart
This item is not returnable. View return policy

Utilize the globally valued, highly accurate, and precise Thermo Scientific™ CEDIA™ Benzodiazepine Assay for very stable shelf life and lot-to-lot dependability.

Utilize the globally valued, highly accurate, and precise Thermo Scientific™ CEDIA™ Benzodiazepine Assay for very stable shelf life and lot-to-lot dependability. Applications are available for an array of clinical chemistry analyzers. The β-Glucuronidase Enzyme, when used with this assay, offers enhanced sensitivity of parent drug by hydrolyzing the glucuronide metabolites. The Thermo Scientific CEDIA panel of drugs of abuse assays is ever expanding to address emerging drug trends and testing needs.

  • One of the largest menus of tests available
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Variety of calibrators and multiconstituent control sets
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Control Sets MGC Multi-Drug Controls
Description CEDIA Benzodiazepine Assays
Detectable Analytes Nitrazepam
DoA Calibrators Multi-drug Calibrator
Sample Type Urine
Type Reagent
Quantity 1 x 495 mL, LC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.