Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CEDIA™ Buprenorphine Drugs of Abuse Calibrators
GSA_VA

Catalog No. 23278211
Change view
Click to view available options
Quantity:
1 x 5 mL
1 x 7.5 mL
Concentration:
0 ng/nL
20 ng/mL
5 ng/mL
50 ng/mL
75 ng/mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
23-278-211 1 x 5 mL 20 ng/mL
23-278-217 1 x 7.5 mL 0 ng/nL
23-278-213 1 x 5 mL 5 ng/mL
23-278-212 1 x 5 mL 50 ng/mL
23-278-214 1 x 5 mL 75 ng/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 23-278-211 Supplier Thermo Scientific™ Supplier No. 100243
Add to Cart
Add to Cart
This item is not returnable. View return policy

Utilize the globally valued accuracy, precision and lot-to-lot dependability of CEDIA™ Buprenorphine Drugs of Abuse Calibrators.

Utilize the globally valued accuracy, precision and lot-to-lot dependability of CEDIA™ Buprenorphine Drugs of Abuse Calibrators, which are optimized for an array of clinical chemistry analyzers. The drugs of abuse testing scope is expanding rapidly, and CEDIA assays meet this challenge by offering one of the most extensive menus for drugs of abuse testing on the market.

  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Applications for an array of clinical chemistry analyzers
  • Variety of calibrators and multiconstituent control sets

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description CEDIA; 20ng/mL; 5mL
Concentration 20 ng/mL
Detectable Analytes Buprenorphine
Sample Type Urine
Type Calibrator
Packaging 1 Bottle
Quantity 1 x 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.