Learn More
Thermo Scientific™ CEDIA™ Cyclosporine (CsA) Immunoassays and Calibrators
Description
Achieve precise and accurate quantitative results crucial for monitoring drug levels of cyclosporine in kidney, liver and heart transplant patients with our easy-to-use Thermo Scientific™ CEDIA™ Cyclosporine Plus Immunoassay. Used with the Thermo Scientific CEDIA Plus High Range Calibrators, this immunoassay achieves a broad dynamic range of 25-2000ng/mL for both trough and peak samples. Applications are available on our easy-to-use Indiko™ and Indiko Plus clinical and specialty chemistry systems as well as a wide variety of other laboratory instrumentation.
- Excellent low-end sensitivity
- Correlation with gold standard methods
- Precise throughput standard curve assay range
- Lot-to-lot consistency
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Detectable Analytes | Cyclosporin A |
| Sample Type | Plasma, Serum, Plasma, Serum |
| Type | CsA Control High Level 5 |
| Quantity | 6 x 2 mL each, 2 x 8 mL diluent |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.