Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CEDIA™ Methadone Metabolite Drugs of Abuse Assays

Catalog No. 23278229
Change view
Click to view available options
Quantity:
Indiko 3 x 17 mL
MCC 65 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-278-229 MCC 65 mL
23-278-241 Indiko 3 x 17 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-278-229 Supplier Thermo Scientific™ Supplier No. 100096
Add to Cart
Add to Cart

Utilize the globally valued accuracy, precision, lot-to-lot dependability and stable shelf life of Thermo Scientific™ CEDIA™ Methadone Metabolite Drugs of Abuse Assays.

Utilize the globally valued accuracy, precision, lot-to-lot dependability and stable shelf life of Thermo Scientific™ CEDIA™ Methadone Metabolite Drugs of Abuse Assays, which offer applications for an array of clinical chemistry analyzers. The drugs of abuse testing scope is expanding rapidly, and CEDIA assays meet this challenge by offering one of the most extensive menus for drugs of abuse testing and developing new assays to respond to ever-emerging drug screening needs.

  • One of the largest menus of tests available
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Variety of calibrators and multiconstituent control sets
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Control Sets MGC Multi-Drug Controls
Detectable Analytes Methadone Metabolite (EDDP)
DoA Calibrators Multi-drug Calibrator
Sample Type Urine
Type Methadone Metabolite Assay
Quantity MCC 65 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.