Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CEDIA™ MultiDrug Drugs of Abuse Calibrators

Catalog No. 23278257
Change view
Click to view available options
Quantity:
1 x 5 mL
1 x 15 mL
Type:
Clinical Calibrator
High Calibrator
Intermediate Calibrator
Primary Calibrator
Secondary Calibrator
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
23-278-257 1 x 5 mL High Calibrator
23-278-258 1 x 5 mL Intermediate Calibrator
23-278-248 1 x 5 mL Clinical Calibrator
23-278-260 1 x 5 mL Secondary Calibrator
23-278-252 1 x 15 mL Secondary Calibrator
23-278-250 1 x 15 mL Intermediate Calibrator
23-278-256 1 x 15 mL High Calibrator
23-278-251 1 x 5 mL Primary Calibrator
23-278-246 1 x 15 mL Primary Calibrator
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 23-278-257 Supplier Thermo Scientific™ Supplier No. 1730398
Add to Cart
Add to Cart
This item is not returnable. View return policy

Utilize the globally valued accuracy, precision and lot-to-lot dependability of CEDIA™ Multidrug Drugs of Abuse Calibrators.

Utilize the globally valued accuracy, precision and lot-to-lot dependability of CEDIA™ Multidrug Drugs of Abuse Calibrators, which are optimized for an array of clinical chemistry analyzers. The drugs of abuse testing scope is expanding rapidly, and CEDIA assays meet this challenge by offering one of the most extensive menus for drugs of abuse testing on the market.

  • Largest menu of tests available
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Applications for an array of clinical chemistry analyzers
  • Variety of calibrators and multiconstituent control sets
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Do not Freeze; 2° to 8°C
Concentration Multianalyte
Detectable Analytes Multiple
Sample Type Urine
Type High Calibrator
Packaging 1 Bottle
Quantity 1 x 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.