Learn More
Thermo Scientific™ CEDIA™ Mycophenolic Acid Immunoassays, Calibrators and Controls
Description
Complete your immunosuppressant drug testing panel with the Thermo Scientific™ CEDIA™ Mycophenolic Acid Immunoassay. Like our other ISD assays, the CEDIA Mycophenolic Acid Immunoassay is accurate and precise, providing users with confidence in their results. Applications are available on our flexible and intuitive Indiko™ and Indiko Plus clinical and specialty chemical systems as well as a wide variety of other laboratory instrumentation.
- Excellent low-end sensitivity
- Correlation with gold standard methods
- Precise throughput standard curve assay range
- Lot-to-lot consistency
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Description | CEDIA Mycophenolic Acid Calibrator Set |
| Detectable Analytes | Mycophenolic Acid |
| Sample Type | Plasma |
| Type | Calibrator |
| Quantity | 5 mL, 2 each level |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.