Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CEDIA™ THC Multi-Level Control Set

Catalog No. 23278282
Change view
Click to view available options
Concentration:
100 ng/mL
25 ng/mL
50 ng/mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration
23-278-282 100 ng/mL
23-278-283 25 ng/mL
23-278-284 50 ng/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 23-278-282 Supplier Thermo Scientific™ Supplier No. 1661060
Add to Cart
Add to Cart
This item is not returnable. View return policy

Eliminate time-consuming steps yet achieve high analytical accuracy with the ready-to-use Thermo Scientific™ CEDIA™ THC (Cannabinoid) Multi-Level Control Set.

Eliminate time-consuming steps yet achieve high analytical accuracy with the ready-to-use Thermo Scientific™ CEDIA™ THC (Cannabinoid) Drugs of Abuse Control Set in liquid form. These assays have been field-tested on a wide range of analyzers, and they are enthusiastically accepted in laboratories where cost control and optimal productivity are key issues.

  • Largest menu of tests available
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Applications for an array of clinical chemistry analyzers
  • Variety of calibrators and multiconstituent control sets
TRUSTED_SUSTAINABILITY

Specifications

Description CEDIA THC 100 Multi-Level Control Set
Concentration 100 ng/mL
Detectable Analytes 11-nor-9-COOH-D9-THC
Sample Type Urine
Type Control
Level Low (75ng/mL) and High (125ng/mL)
Quantity 1 x 15 mL Low and 1 x 15 mL High
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.