Learn More
Thermo Scientific™ CEDIA™ Therapeutic Drug Monitoring (TDM) Assays
Description
Receive reliable performance on a wide range of analyzers with Thermo Scientific™ CEDIA™ Therapeutic Drug Monitoring (TDM) Assays. Manufactured since 1986, they provide an excellent range of Thermo Scientific™ DRI™ TDM Assays as well as CEDIA TDM Assays and Calibrators.
- Wide dynamic measuring range of analytes as low as 0.15ng/mL for Digoxin
- Extended linearity for analytes like Cyclosprine at 25ng/mL to 2000ng/mL range
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Description | Phenytoin II Assay |
| Detectable Analytes | Phenytoin |
| Sample Type | Plasma, Serum |
| Type | Reagent |
| Quantity | 17 x 17 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.