Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CEDIA™ Therapeutic Drug Monitoring (TDM) Assays

Catalog No. 23278275
Change view
Click to view available options
Description:
Carbamazepine Assay
Phenobarbital II Assay
Phenytoin II Assay
Procainamide Assay
Theophylline II Assay
Valproic Acid Assay
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
23-278-275 Valproic Acid Assay
23-278-286 Carbamazepine Assay
23-278-232 Phenobarbital II Assay
23-278-233 Phenytoin II Assay
23-278-288 Procainamide Assay
23-278-235 Theophylline II Assay
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 23-278-275 Supplier Thermo Scientific™ Supplier No. 100013
Add to Cart
Add to Cart

Receive reliable performance on a wide range of analyzers with Thermo Scientific™ CEDIA™ Therapeutic Drug Monitoring (TDM) Assays.

Receive reliable performance on a wide range of analyzers with Thermo Scientific™ CEDIA™ Therapeutic Drug Monitoring (TDM) Assays. Manufactured since 1986, they provide an excellent range of Thermo Scientific™ DRI™ TDM Assays as well as CEDIA TDM Assays and Calibrators.

  • Wide dynamic measuring range of analytes as low as 0.15ng/mL for Digoxin
  • Extended linearity for analytes like Cyclosprine at 25ng/mL to 2000ng/mL range
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description Valproic Acid Assay
Detectable Analytes Valproic Acid
Sample Type Plasma, Serum
Type Reagent
Quantity 13 x 11 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.