Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cediranib maleate, Thermo Scientific™

Catalog No. AC468952500
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC468952500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC468952500 Supplier Thermo Scientific Chemicals Supplier No. 468952500
Add to Cart

Specifications

Chemical Name or Material Cediranib maleate
CAS 857036-77-2
Quantity 250 mg
Shipping Condition No special requirements
Product Line Life science
Molecular Formula C25 H27 F N4 O3 . C4 H4 O4
Physical Form Powder
Color White to Beige
Purity ≥97.5%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.