Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cefamandole nafate

Click to view available options
Quantity:
5g
10g
42540-40-9
C19H17N6NaO6S2
512.491
MFCD00864877
ICZOIXFFVKYXOM-YCLOEFEOSA-M
23665731
CHEBI:3481
sodium;(6R,7R)-7-[[(2R)-2-formyloxy-2-phenylacetyl]amino]-3-[(1-methyltetrazol-5-yl)sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
CN1C(=NN=N1)SCC2=C(N3C(C(C3=O)NC(=O)C(C4=CC=CC=C4)OC=O)SC2)C(=O)[O-].[Na+]
42540-40-9
C19H17N6NaO6S2
512.491
MFCD00864877
ICZOIXFFVKYXOM-YCLOEFEOSA-M
23665731
CHEBI:3481
sodium;(6R,7R)-7-[[(2R)-2-formyloxy-2-phenylacetyl]amino]-3-[(1-methyltetrazol-5-yl)sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
CN1C(=NN=N1)SCC2=C(N3C(C(C3=O)NC(=O)C(C4=CC=CC=C4)OC=O)SC2)C(=O)[O-].[Na+]

A second generation cephalosporin antibiotic

Solubility Information Freely soluble in water; Sparingly soluble in methanol
Formula Weight 513.5
Physical Form Powder
Color White
Melting Point 190°C
Quantity 10g
Chemical Name or Material Cefamandole nafate

GHS H Statement
H334-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

GHS P Statement
P285-P261-P280-P305+P351+P338-P405-P501a
In case of inadequate ventilation wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : XI0380000

storageNote1 : 2° to 8°C