Learn More
Description
- Ready to use
- No preparation or dilution of antimicrobial agent required
- Place on an inoculated agar plate and incubate
- MIC testing for a wide range of agents and against many organisms can be done on request and at short notice
- Synergy Testing
- Can be crossed for evaluating a critical assay such as the in vitro combination of antibiotics,
- Rapid and efficient
- Full range of MIC testing of 6 agents on a large plate takes no more than 5 min.
- Solid Medium
- Growth of organisms on agar better mimics in vitro conditions
- Easy to visualize hetero-resistant variants in population and detect mixed cultures and contamination
- Package small and lightweight
- Wide MIC range
- Spans across 15 dilutions fulfilling pharmacokinetic needs
- Varying susceptibility of many organism groups, covers different MIC breakpoints used in different countries as recommended by various reference or expert groups
- Easy to learn
- Minimal training required for clinical microbiology personnel
- No equipment is required for use
- Easy for new agents
- MIC equivalent to reference values
- Improved MIC precision
- Flexible media choice
- Can be used as a quality control tool for checking different AST media, brand to brand and batch to batch
- Robust technique
- Generates quantitative MIC values that needed to guide therapy selection for individual patient
Specifications
Specifications
| Clia Complexity | Not Applicable |
| Detectable Analytes | Cefepime-taniborbactam |
| Type | Test Strips |
| Disposable | Single-use |
| For Use With (Application) | MIC Testing |
| Quantity | 10 Pk. |
| Form | Ready-to-Use (RTU) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
