Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cefixime (trihydrate) | 125110-14-7 | 99.8% | 507.50 g·mol⁻¹ | C16H21N5O10S2 | 10 MG
SDP

Supplier:  Medchemexpress LLC HYB1381A10MG

Encompass_Preferred

Cefixime trihydrate is a third-generation cephalosporin antibiotic used as a research reagent and analytical standard for studying antibacterial activity and for analytical testing.

  • Third-generation cephalosporin antibiotic.
  • High purity analytical standard (COA example 99.77%).
  • White to off-white solid, easily handled in laboratory settings.
  • Molecular weight 507.50 g·mol⁻¹; molecular formula C16H21N5O10S2.
  • CAS number 125110-14-7 for unambiguous identification.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-97798


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.