Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cefixime Trihydrate, G-Biosciences
SCI_ED

Catalog No. S000005
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000005 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S000005 Supplier G-Biosciences Supplier No. RC922
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cefixime trihydrate is a broad-spectrum third generation cephalosporin targeting a wide range of gram-positive and gram-negative organisms.

Antibiotics are often used in clinical in vitro tests known as antimicrobial susceptibility tests to determine their efficacy against certain bacterial species.

Chemical Identifiers

Molecular Weight (g/mol) 507.50 g/mol

Specifications

Quantity 100 mg
Percent Purity 98%
Grade Molecular Biology
Physical Form Powder
Product Line Gibco
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.