Learn More
TARGETMOL CHEMICALS INC Cefotaxime sodium 500MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T0911500MG
Encompass_Preferred
Also available in 1 mL, 100 mg, 250 mg and bulk. Please contact Fisher for quotes. Cefotaxime is a third-generation cephalosporin antibiotic. Like other third-generation cephalosporins, it has broad spectrum activity against Gram positive and Gram negative bacteria. In most cases, it is considered to be equivalent to ceftriaxone in terms of safety and efficacy. Cefotaxime sodium (Cefotaxim sodium salt) is marketed under various trade names including Claforan (Sanofi-Aventis). Purity 99.51%
Catalog No. 50-288-0726
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.