Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cefotaxime Sodium Salt, ≥95%, MP Biomedicals™

Catalog No. ICN15494790 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
0.1 g
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15494790 500 mg
ICN15494780 0.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15494790 Supplier MP Biomedicals, Inc Supplier No. 0215494790
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 64485-93-4 | Formula: C16H16N5NaO7S2 | Weight: 477.44 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64485-93-4
Molecular Formula C16H16N5NaO7S2
Molecular Weight (g/mol) 477.44
MDL Number MFCD00079073
InChI Key AZZMGZXNTDTSME-JUZDKLSSSA-M
Synonym (6R-(6-α,7-β(Z)))-3-((Acetyloxy) methyl)-7- (((2-amino-4-thiazolyl) (methoxyimio) acetyl) amino)-8- oxo-5-thia-1- azabicyclo (4,2,0) oct-2-ene-2-carboxylic acid, sodium salt, (6R-(6-α, 7-β(Z)))-3-((Acetyloxy) methyl)-7- (((2-amino-4-thiazolyl) (methoxyimio) acetyl) amino)-8- oxo-5-thia-1- azabicyclo (4, 2
PubChem CID 88631411
SMILES [Na+].CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(COC(C)=O)=C(N2C1=O)C([O-])=O)C1=CSC(N)=N1

Specifications

Type Cefotaxime Sodium Salt
Physical Form Powder
Color White
Quantity 500 mg
Assay Percent Range ≥95%
IUPAC Name sodium;(6S,7R)-3-(acetyloxymethyl)-7-[[2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
Formula Weight 477.5
Percent Purity 95%
Grade Not Available
Chemical Name or Material Cefotaxime Sodium Salt
EINECSNumber 264-915-9
Recommended Storage Store at + 4°C, protect from light.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.