CAS RN 64485-93-4
Cefotaxime sodium salt
CAS: 64485-93-4 Molecular Formula: C16H16N5NaO7S2 Molecular Weight (g/mol): 477.44 MDL Number: MFCD00079073 InChI Key: AZZMGZXNTDTSME-JUZDKLSSSA-M PubChem CID: 88631411 IUPAC Name: sodium (6R,7R)-3-[(acetyloxy)methyl]-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetamido]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate SMILES: [Na+].CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(COC(C)=O)=C(N2C1=O)C([O-])=O)C1=CSC(N)=N1
Cefotaxime Sodium Salt 96.0+%, TCI America™
CAS: 64485-93-4 Molecular Formula: C16H16N5NaO7S2 Molecular Weight (g/mol): 477.44 MDL Number: MFCD00079073 InChI Key: AZZMGZXNTDTSME-JUZDKLSSSA-M Synonym: cefotaxime sodium,sodium 6r,7r-3-acetyloxy methyl-7-2-2-amino-1,3-thiazol-4-yl-2-methoxyimino acetamido-8-oxo-5-thia-1-azabicyclo 4.2.0 oct-2-ene-2-carboxylate,cefoperazone sodium salt u.s.p.,6r-6-a,7-b z-3-acetyloxy methyl-7-2-amino-4-thiazolyl methoxyimio acetyl amino-8-oxo-5-thia-1-azabicyclo 4,2,0 oct-2-ene-2-carboxylic acid, sodium salt,sodium 3-acetyloxymethyl-7-2-2-amino-4-thiazolyl-2-methoxyimino-1-oxoethyl amino-8-oxo-5-thia-1-azabicyclo 4.2.0 oct-2-ene-2-carboxylate,sodium 3-acetyloxymethyl-7-2-2-azanyl-1,3-thiazol-4-yl-2-methoxyimino-ethanoyl amino-8-oxidanylidene-5-thia-1-azabicyclo 4.2.0 oct-2-ene-2-carboxylate,sodium 3-acetyloxy methyl-7-2-amino-1,3-thiazol-4-yl methoxyimino acetyl amino-8-oxo-5-thia-1-azabicyclo 4.2.0 oct-2 PubChem CID: 88631411 IUPAC Name: sodium (6R,7R)-3-[(acetyloxy)methyl]-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetamido]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate SMILES: [Na+].CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(COC(C)=O)=C(N2C1=O)C([O-])=O)C1=CSC(N)=N1
Thermo Scientific Chemicals Cefotaxime sodium salt, 95%
CAS: 64485-93-4 Molecular Formula: C16H16N5NaO7S2 Molecular Weight (g/mol): 477.44 MDL Number: MFCD00079073 InChI Key: AZZMGZXNTDTSME-JUZDKLSSSA-M PubChem CID: 88631411 SMILES: [Na+].CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(COC(C)=O)=C(N2C1=O)C([O-])=O)C1=CSC(N)=N1
Cefotaxime Sodium Salt, ≥95%, MP Biomedicals™
CAS: 64485-93-4 Molecular Formula: C16H16N5NaO7S2 Molecular Weight (g/mol): 477.44 MDL Number: MFCD00079073 InChI Key: AZZMGZXNTDTSME-JUZDKLSSSA-M Synonym: (6R-(6-α,7-β(Z)))-3-((Acetyloxy) methyl)-7- (((2-amino-4-thiazolyl) (methoxyimio) acetyl) amino)-8- oxo-5-thia-1- azabicyclo (4,2,0) oct-2-ene-2-carboxylic acid, sodium salt,(6R-(6-α, 7-β(Z)))-3-((Acetyloxy) methyl)-7- (((2-amino-4-thiazolyl) (methoxyimio) acetyl) amino)-8- oxo-5-thia-1- azabicyclo (4, 2 PubChem CID: 88631411 SMILES: [Na+].CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(COC(C)=O)=C(N2C1=O)C([O-])=O)C1=CSC(N)=N1
Cefotaxime Sodium Salt, MP Biomedicals™
CAS: 64485-93-4 Molecular Formula: C16H16N5NaO7S2 Molecular Weight (g/mol): 477.44 MDL Number: MFCD00079073 InChI Key: AZZMGZXNTDTSME-JUZDKLSSSA-M Synonym: Cefotaxime sodium, (6R-(6-a,7-b(Z)))-3-((Acetyloxy) methyl)-7- (((2-amino-4-thiazolyl) (methoxyimio) acetyl) amino)-8- oxo-5-thia-1- azabicyclo (4,2,0) oct-2-ene-2-carboxylic acid, sodium salt PubChem CID: 88631411 IUPAC Name: sodium;(6S,7R)-3-(acetyloxymethyl)-7-[[2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate SMILES: [Na+].CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(COC(C)=O)=C(N2C1=O)C([O-])=O)C1=CSC(N)=N1