Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cefotiam HCl, G-Biosciences
SCI_ED

Catalog No. S903432
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S903432 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S903432 Supplier G-Biosciences Supplier No. RC934
Add to Cart
Add to Cart

Cefotiam is a second generation cephalosporin antibiotic effective against gram-negative and gram-positive bacteria. However, it is not effective against Pseudomonas aeruginosa.

Cefotiam has been used as a prophylactic antibiotic against methicillin-resistant Staphylococcus aureus or MRSA.

Chemical Identifiers

Molecular Weight (g/mol) 562.09 g/mol

Specifications

Boiling Point 94°C
Quantity 100 mg
Percent Purity 98%
Grade Laboratory
Physical Form Powder
Product Line Gibco
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.