Learn More
Cefoxitin sodium salt, Analytical Standard, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| Packaging | Glass Bottle |
| CAS | 33564-30-6 |
| Physical Form | Neat |
| Linear Formula | C16H16N3NaO7S2 |
| MDL Number | MFCD00079042 |
| Quantity | 1 g |
| Formula Weight | 449.43 |
| Grade | Analytical Standard |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.