Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cefozopran HCl, G-Biosciences
SCI_ED

Catalog No. S960101
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S960101 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S960101 Supplier G-Biosciences Supplier No. RC936
Add to Cart
Add to Cart

Cefozopran is a fourth generation cephalosporin antibiotic effective against a variety of gram-negative and gram-positive organisms, including Pseudomonas aeruginosa.

It is effective against MRSA when combined with antibiotics such as vancomycin and teicoplanin.

Chemical Identifiers

Molecular Weight (g/mol) 551.99 g/mol

Specifications

Quantity 100 mg
Percent Purity 98%
Grade Molecular Biology
Physical Form Powder
Product Line Gibco
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.