Learn More
AdipoGen Cefpodoxime
SDP
Supplier:  AdipoGen CDXC0361M025
Encompass_Preferred
Chemical. CAS 80210-62-4. Formula C15H17N5O6S2. MW 427.46. Synthetic. Cefpodoxime is a metabolite of cefpodoxime proxetil, an oral third generation cephalosporin antibiotic. It is active against most Gram positive and Gram negative bacteria MIC90 ranges from 0.015-8 µg/ml. It is commonly used to treat acute otitis media, pharyngitis, and sinusitis. Cefpodoxime inhibits cell wall synthesis by inhibiting the final transpeptidation step of peptidoglycan synthesis in cell walls.
Catalog No. 50-256-1365
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.