Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cefsulodin (SCE-129), G-Biosciences
SCI_ED

Catalog No. S000008
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000008 5 g
S000007 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S000008 Supplier G-Biosciences Supplier No. RC946
Add to Cart
Add to Cart

Cefsulodin is a third generation cephalosporin antibiotic used as a selective agent in isolation media such as Yersinia Selective Agar, Columbia Blood Agar, Salmonella Chromogenic Agar, and CIN Agar.

It is mildly effective against gram-positive and gram-negative bacteria, but exhibits greater efficacy against Pseudomonas aeruginosa.

Chemical Identifiers

Molecular Weight (g/mol) 554.53 g/mol

Specifications

Quantity 5 g
Percent Purity >95%
Grade Molecular Biology
Physical Form Powder
Product Line Gibco
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.