Learn More
Description
- Discs impregnated with antibiotic
- Used for susceptibility test according to Kirby-Bauer antibiotic testing (KB testing or disc diffusion antibiotic sensitivity testing)
- Antibiotic discs clearly marked on both sides with letters and numbers designating agent and drug content
- In vitro semi-quantitative method for antimicrobial susceptibility of clinical isolates tested on agar media using overnight incubation
- 250-test box contains 5 cartridges of 50 discs
- Each cartridge individually packed in desiccant envelope
Specifications
Specifications
| Format | Paper Disc |
| Product Type | Antimicrobial Susceptibility Disc |
| Target Organism Class | Gram-negative Microorganisms, Gram-positive Microorganisms, Enterobacterales |
| Test Type | Antimicrobial Susceptibility |
| Species | Staphylococcus aureus |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
