Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Ceftriaxone Disodium Salt Hemi(heptahydrate), Beta-lactam Antibiotic, 50 mg

Catalog No. 502707182
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-7182 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-7182 Supplier Abcam Supplier No. AB12087150MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity beta-lactam antibiotic compound for research applications.

  • Molecular Weight: 1323.2 Da
  • Purity: >98%
  • Form: Solid
  • Constituents: 100% Ceftriaxone disodium salt hemi(heptahydrate)
  • CAS Number: 104376-79-6
  • Storage Temperature: Store at +4°C
  • Shipping Temperature: Ambient - Can Ship with Ice

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Content And Storage Store at 4°C.
Buffer 100% Ceftriaxone disodium salt hemi(heptahydrate)
Form Solid
Shipping Condition Ambient
Target Molecule Ceftriaxone
Formulation 100% Ceftriaxone disodium salt hemi(heptahydrate)
Quantity 50 mg
For Use With (Application) Antibiotic research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.