Learn More
Grainger Ceiling Strobe Marked Fire White
Supplier:  Grainger 16X394
Encompass_Preferred
The Edwards Signaling 24V DC Strobe Light is a weatherproof strobe light designed for outdoor use. It has a polycarbonate lens and housing and operates on 24V DC. The light output is rated at 75 candela and it flashes at 1Hz. Some key features include a universal mounting bracket, plug-in terminal block wiring, and it meets UL 1638. This strobe light can be used for alarm signaling, emergency notification, or other signaling applications requiring a weatherproof and durable strobe.
Catalog No. 50-276-6317
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.