Learn More
New Pig Ceiling Tile Leak Diverter
SDP
Catalog No. 18389151 Shop All New Pig Corporation Products
Change view
Click to view available options
Type: Ceiling Tile Leak Diverter
Catalog No.
18-389-151 Type: Ceiling Tile Leak Diverter
1 options
This item is not returnable. View return policy
Catalog No. 18-389-151 Supplier New Pig Supplier No. TLS367
Add to Cart
Add to Cart
This item is not returnable. View return policy
Leak diversion system replaces damaged ceiling tile for discreet channeling of liquids until you find and fix the leak.
  • Remove damaged tile and replace it with a leak diverter panel Easily keep rain and snow from storming inside
  • Whether condensation and ice, or heavy rain from a high-wind hurricane, this solution will quickly protect your team and what you've built
  • Captures leaks and drips and diverts them to a drain or collection container with included 25'L hose
  • Clips let you secure hose to ceiling grids for a professional appearance
  • After leak has been repaired, simply uninstall and store for later use Replaces any standard 2' x 4' drop ceiling tile
  • Flow rate of 0.2 gallons per minute when used with included hose
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov