Learn More
Enzo Life Sciences Celastrol (5mg). CAS: 34157-83-0
SDP
Supplier:  Enzo Life Sciences ALX350332M005
Encompass_Preferred
Cell permeable triterpenoid; anti-inflammatory and immunosuppressive compound. Antioxidant. Suppresses LPS-induced cytokine release in macrophages and monocytes (IC50=40nM for IL-1β and IL-1α, 80nM for IL-6, 110nM for prostaglandin E2 and 210nM for IL-8 and TNF-α). Suppresses nitric oxide (NO) production (IC50=230nM) and LPS-induced NF-κB activation (IC50=270nM). Inhibits chymotrypsin-like activity of 20S proteasome (IC50=2.5µM). Inhibits lipid peroxidation induced by ADP and Fe2+ in rat liver mitochondria (IC50=7µM). Induces expression of a wider set of heat shock proteins. Inhibits topoisomerase II. Alternative name: Tripterine, 3-Hydroxy-24-nor-2-oxo-1(10),3,5,7-friedelatetraen-29-oic acid. Purity: ≥98% (HPLC). Solubility: Soluble in 100% ethanol, DMSO or dimethyl formamide; insoluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-8845
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.