Learn More
Celatom™ Acid-washed for use in Total Dietary Fiber Assay, TDF-100A, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsDescription
Specifications
Specifications
| CAS | 68855-54-9 |
| Quantity | 1 kg |
| MDL Number | MFCD00132803 |
| Packaging | Poly Bottle |
| Product Line | Celatom |
Celatom is a registered trademark of EP Minerals, LLC
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.