Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Celestial Star Globe
SDPSCI_ED

Catalog No. S89562 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
5 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S89562 5 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy

Prop 65 Warning: Cancer and reproductive harm

Catalog No. S89562 Supplier United Scientific™ Supplier No. CELEST1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Prop 65 Warning: Cancer and reproductive harm

United Scientific™ Celestial Star Globe is a demonstration device that shows the apparent positions of the stars visible from Earth.

The Celestial Globe is a demonstration device that shows the apparent positions of the stars visible from Earth. The three dimensional representation allows the relative angular positions of the constellations to be visualized and permits students to estimate which stars will be visible from different places on Earth for any rotational position of the planet.

  • Consists of two concentric spheres mounted on a common axle and supported on a frame with a base.
  • The inner sphere (1 in the above illustration) is 4 in. in diameter and represents the Earth. The continents are marked on it in their correct relative positions.
  • The outer sphere (2) is transparent. It is 12.5 in. in diameter and has the constellations and their stars marked on it. Yellow shading indicates the Milky Way, the plane of our galaxy.
  • The axle (3) that carries both spheres allows them to be rotated independently.
  • An angle scale is marked on the frame (4) that supports the axle. The angles are marked every degree and extend from zero at the equator to 87° in both directions.
  • The frame is fixed to a round base (5) so that the axle is held at an angle of 23.5° to the vertical, representing the inclination of the Earth's spin axis to the axis of its orbit around the sun.

Specifications

Grade 5 to 12
Format Kit
Product Type Celestial Globe
Languages English
For Use With (Application) Estimate Which Stars Will Be Visible From Different Places On Earth
Subject Geology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.