Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Urea, 98%, extra pure, pearls

Catalog No. AC197460100
Click to view available options
Quantity:
1 kg
5 kg
10 kg
Packaging:
Plastic Bottle
Plastic drum

CAS: 57-13-6 | CH4N2O | 60.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-13-6
Molecular Formula CH4N2O
Molecular Weight (g/mol) 60.06
MDL Number MFCD00008022
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
Synonym carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
PubChem CID 1176
ChEBI CHEBI:48376
IUPAC Name urea
SMILES C(=O)(N)N

Specifications

Melting Point 131.0°C to 135.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ex N)
Packaging Plastic drum
Linear Formula H2NCONH2
Quantity 10 kg
Beilstein 03, 42
Fieser 01,1262; 02,455
Merck Index 15, 10052
Solubility Information Solubility in water: 1080g/L (20°C). Other solubilities: soluble in alcohol, benzene, concentrated hcl, practically insoluble in chloroform and ether, soluble in methanol: 16.6 g/100 mL, soluble in ethanol: 5 g/100 mL
Formula Weight 60.06
Percent Purity 98%
Grade Extra Pure
Physical Form Pearls
Chemical Name or Material Urea, 98%
Show More Show Less
Hazard Statement GHS H Statement:
Causes skin irritation.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-315-5
RTECSNumber YR6250000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.