Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ Cell Culture Dishes, Non-Treated

Catalog No. 13690078 Shop All Eppendorf North America Products
Change view
Click to view available options
Diameter (Metric):
40.8 mm
Culture Area:
9.5 sq. cm
Volume (Metric) Working:
2 to 3 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Culture Area Volume (Metric) Working
13-690-078 40.8 mm 9.5 sq. cm 2 to 3 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 13-690-078 Supplier Eppendorf™ Supplier No. 0030700015
This item is not returnable. View return policy

For cell based assays and microscopic analysis. Eppendorf™ Cell Culture Dishes, Non-Treated helps in direct access to the cells.

  • Safety in handling during transportation and while working due to corrugated handling ring
  • SplashProtect ring inside of the dish lid traps liquid and prevents spills during transportation or incubation
  • Pronounced rim on dish lid and exact fitting of lid and bottom ensure sturdy, secure stacking
  • Easy differentiation of lid and dish bottom prevents unintended removal of the lid
  • Packaging combines tool-free opening, resealable top and side for tape-free closing, and an easy solution to shrink the package for space-conscious storage
  • Available in three formats: 35, 60 and 100mm
TRUSTED_SUSTAINABILITY

Specifications

Closure Color Transparent
Closure Included Closure Included
Closure Material Polystyrene
Autoclavable Not Autoclavable
Bottom Shape Flat
Capacity (Metric) 8.1 mL
Color Transparent
Culture Area 9.5 sq. cm
Material Polystyrene
Shape Round
Sterility Sterile
Surface Treatment Non-treated
Type Cell Culture Dish
Volume (Metric) Working 2 to 3 mL
Includes Lid
Height (Metric) 10.8 mm
Diameter (Metric) 40.8 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.