Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ BRAND™ cell-culture™ Hand Controller

Catalog No. 01670231 Shop All BRANDTECH Scientific Products
Change view
Click to view available options
For Use With (Equipment):
QuikSip BT-Aspirator
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
01-670-231 QuikSip BT-Aspirator
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 01-670-231 Supplier BRANDTECH™ BRAND™ Supplier No. 25950
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Aspiration Controller for use with Pasteur pipets, pipette tips, or QuikSip™ bottle-top aspirator.

  • The cell-culture™ aspiration controller facilitates the removal of surplus liquids.
  • Comes equipped with three interchangeable adapters designed to accommodate pasteur pipettes, capillaries, or pipette tips (up to 100 μL).
  • Adapter can be rotated for effortless fitting of pipettes or pipette tips
  • To aspirate liquids, the instrument must be connected to both a collection bottle and a vacuum pump, or as an alternative to a vacuum pump, the QuikSip™ BT-Aspirator can be used instead (all sold separately)
  • The cell-culture™ aspiration controller is not autoclavable, but the included tube and adapters are.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Aspiration Controller
Autoclavable Autoclavable
Color White
Channels 1
Material Silicone
For Use With (Application) Fluid Aspiration
For Use With (Equipment) QuikSip BT-Aspirator
Includes Suction Tube, Three Adapters
Sterility Non-sterile
Warranty 1 Year
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.