Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ Cell Culture Plates, Non-Treated

Catalog No. 13690069 Shop All Eppendorf North America Products
Change view
Click to view available options
Diameter (Metric):
22.4 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
13-690-069 22.4 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 13-690-069 Supplier Eppendorf™ Supplier No. 0030721012

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Tailored for expansion of smaller cell numbers as well as for cell-based assays. Eppendorf™ Cell Culture Plates, Non-Treated helps in monitoring qualification of cell morphology and cell performance.

  • Easy and fast well identification by contrast rich individual well ID and OptiTrack alphanumeric labeling
  • Moat surrounding outer wells to prevent the “edge effect” when filled with liquid
  • Chimney-well design enables filling of inter-well spaces of plate to level out well-to-well temperature shifts outside incubator
  • Robust stacking performance by pronounced rims on plate lid
  • Lid taps under plate for minimized surface contact and reduce risk of contamination
  • Easy differentiation of lid and plate bottom due to a pronounced corrugation and wider sized base
  • Pronounced ventilation gaps for optimized gas and temperature transfer when incubating in stacks
  • Optimized microscopical performance due to enhanced planarity, reduction of meniscus and clarity of the material
  • Available in 6, 12, 24, 48 and 96-Well format
TRUSTED_SUSTAINABILITY

Specifications

Array 3 x 4
Closure Color Transparent
Closure Included Closure Included
Closure Material Polystyrene
Autoclavable Not Autoclavable
Bottom Shape Flat
Capacity (Metric) 6.8 mL
Color Transparent
Culture Area 391.1 sq. mm
Material Polystyrene
Shape Rectangular
Sterility Sterile
Surface Treatment Non-treated
Type Cell Culture Dish
Volume (Metric) Working 1 to 2 mL
Well Count 12
Dnase Free Yes
Includes Lid
Quantity 60 Plates
RNase Free Yes
Length (Metric) 127.8 mm
Width (Metric) 85.5 mm
Height (Metric) 20 mm
Diameter (Metric) 22.4 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.