Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Cell Cycle and DNA Repair Antibody Pack
SDP

Catalog No. NBP178941 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
3 Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP178941 3 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP178941 Supplier Novus Biologicals™ Supplier No. NBP178941

For Research applications

Specifications

Antigen Cell Cycle and DNA Repair
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunoassay
Conjugate Unconjugated
Dilution Western Blot, Simple Western, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Knockout Validated
Formulation See individual datasheets.
Immunogen See individual datasheets.
Quantity 3 Vials
Research Discipline Cell Cycle and Replication, DNA Repair
Test Specificity This Cell Cycle and DNA Repair Antibodies Sample Pack includes antibodies to Mre11, NBS1 and RAD50.
Includes 1.NB100-142: Mre11 Antibody
2.NB100-143: Nbs1 Antibody
3.NB100-1487: Rad50 Antibody
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.